
Iron(III) sulphate hydrate ≥73.0%, granular, AR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:15244-10-7
:
Formula:Fe₂(SO₄)₃·xH₂O
MDL Number:MFCD00149714
:399.88 g/mol (anhy)
Merck Index:13,04065
Density:3,1 g/cm³ (20 °C)
:Management of Change (MOC) category = N
EINECS:233-072-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:—
- Assay (Fe₂(SO₄)₃):≥73.0 %
- Solubility:Passes Test
- Insoluble Matter:≤0.020 %
- Chloride (Cl):≤0.005 %
- Ferrous Iron:≤0.02 %
- Nitrate (NO₃):≤0.01 %
- Substances not Precipitated by NH₄OH:≤0.10 %
- Copper (Cu):≤0.005 %
- Zinc (Zn):≤0.005 %
- Appearance (tan to light greenish-yellow. Granular or granular powder.):—
Specification Test Results
Frequently Bought Together