

Sodium hydroxide 25% (w/w) in aqueous solution, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Flash Pt:1,27 g/cm³
UN:1824
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:24.50 - 25.50%
- Protect from air to avoid absorption of carbon dioxide.:≤5 ppm
- Assay (NaOH):24.50 - 25.50 %
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
Frequently Bought Together