Order Entry
India
ContactUsLinkComponent
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Flammable
Corrosive
Irritant
Health
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5644-02
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Potassium hydroxide 0.5 N in ethanol, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5644-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.495 - 0.505
  • Normality:
    0.495 - 0.505
  • Appearance:
    Passes Test

Specification Test Results

About this item

 :
 :56.11 g/mol
Density:0,79 g/cm³
Flash Pt:13,00 °C
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553