Order Entry
India
ContactUsLinkComponent
 
 
 
Ethylene glycol ≥99.0% (by GC), AR®, Macron Fine Chemicals™
    
 
undefined
 
 
Ethylene glycol ≥99.0% (by GC), AR®, Macron Fine Chemicals™
    
 

 

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

 

 

  • Acidity (as CH₃COOH):
    ≤ 0.010 %
  • Assay (HO(CH₂)₂OH) (by GC):
    ≥ 99.0 %
  • Chloride (Cl):
    ≤ 0.0005 %
  • Color (APHA):
    ≤ 15
  • Iron (Fe):
    ≤ 0.00002 %
  • Residue after Ignition:
    ≤ 0.005 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤ 0.20 %
  • Specific Gravity at 25°/25°C:
    1.112 - 1.115
  • Boiling Range(Lower):
    193 - 205 °C
  • Boiling Range(Upper):
    193 - 205 °C
  • Appearance (clear, colorless hygroscopic liquid):
    Clear, Colourless Hygroscopic Liquid

 

 

Frequently Bought Together