Order Entry
India
Orders LinkContactUsLinkComponent
AppliClear water

AppliClear water

AppliClear-water
Catalog # A7004.0250
Supplier:   PanReac AppliChem
AppliClear water
Catalog # A7004.0250
Supplier:  PanReac AppliChem
AppliClear water
AppliClear water

AppliClear-water

Price
Inquire For Price
Log in for availability
1 * 250 mL
Product Details & Documents

Biocide is a quaternary ammonium compound.

  • Concentrate for the prevention of growth of algae and moulds in water baths
  • Non toxic
  • With colour indicator (blue to colourless)
  • 1 ml sufficient for 1 L
  • Germ-killing effect on microorganisms

AppliClear-water is fully active, when the colour indicator is blue. When colour turns to colourless, then refill with AppliClear-water.

Caution: Use biocides safely. Always read the label and product information before use.

Specifications
  • Description:
    AppliClear-water
  • Seq:
    02