Order Entry
India
ContactUsLinkComponent
Hydrogen peroxide 3% in aqueous solution stabilised, BAKER ANALYZED®, J.T.Baker®
Hydrogen peroxide 3% in aqueous solution stabilised, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Hydrogen peroxide 3% in aqueous solution stabilised, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    3.0 - 3.5%
  • Assay (H₂O₂):
    3.0 - 3.5 %
  • Residue after Evaporation:
    ≤0.0500 %
  • Free Acid (as H₂SO₄):
    ≤0.01 %
  • Phosphate (PO₄):
    ≤0.003 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Preservative (acetophenetidin):
    ≤0.05 %
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - Arsenic (As):
    ≤0.100 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

Frequently Bought Together