Pyridine ≥99.0% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:110-86-1
:
:C₅H₅N
:MFCD00011732
:13,08060
:115 °C (1013 hPa)
:–44 °C
:0,98 g/cm³
:17 °C
:Ambient
:1282
:3,II
:79.1 g/mol
:203-809-9
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤20
- Assay (C₅H₅N) (by GC, corrected for water):≥99.0 %
- Color (APHA):≤20
- Solubility in H₂O:Passes Test
- Residue after Evaporation:≤0.0020 %
- Water (by KF, volumetric):≤0.1 %
- Sulfate (SO₄):≤0.001 %
- Ammonia (as NH₃):≤0.002 %
- Reducing Substances:Passes Test
- Chloride (Cl):≤5 ppm
- Trace Impurities - Copper (Cu):≤5 ppm
Frequently Bought Together