Order Entry
India
ContactUsLinkComponent
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Corrosive
Irritant
Health
Environment
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Environment
Silver nitrate ≥99.7%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.7%
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Assay (AgNO₃):
    ≥99.7 %
  • Clarity of Solution:
    Passes Test
  • Free Acid:
    Passes Test
  • Substances Not Precipitated by HCl:
    ≤0.01 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Lead (Pb):
    ≤0.001 %
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - Copper (Cu):
    ≤2.000 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm

Specification Test Results

Related Information

Frequently Bought Together