tetra-Chloroauric(III) acid trihydrate, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate
:16961-25-4
:
:HAuCl₄·3H₂O
:MFCD00149904
:393.83 g/mol
:3,90 g/cm³
:3260
:8,II
:GHS05
:GHS07
- Environmentally Preferable:
- Meets ACS Reagent Chemical Requirements:≥49.0%
- For Laboratory,Research,or Manufacturing Use:≤0.1%
- Material is very hygroscopic. Store in a cool place.:≤0.2%
- Assay (as Au):≥49.0 %
- Insoluble in Ether:≤0.1 %
- Alkalies and Other Metals (as SO₄):≤0.2 %
Frequently Bought Together