Magnesium sulphate heptahydrate Ph. Eur., USP, BP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10034-99-8
:
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2,66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-298-2
Specification Test Results
- Appearance:Conforms (see CoA/CoS)
- Heavy metals (as Pb):≤ 10 ppm
- Residual solvents:Passes test
- Residual metal catalysts (skip lot)::.
- As (Arsenic):≤ 2 ppm
- Cd (Cadmium):≤ 0.1 ppm
- Co (Cobalt):≤ 1 ppm
- Hg (Mercury):≤ 2 ppm
- Li:Max. 0.5 ppm
- Ni (Nickel):≤ 6 ppm
- Pb (Lead):≤ 2 ppm
- V (Vanadium):≤ 10 ppm
- Conforms to Ph.Eur./USP/BP:Passes test
- Ph.Eur./BP specifications::.
- Assay (dried substance) [Ph.Eur./BP]:99.0 - 100.5 %
- Identification A:Passes test Ph.Eur./BP
- Identification B:Passes test Ph.Eur./BP
- Appearance of solution:Passes test Ph.Eur./BP
- Acidity or alkalinity:Passes test Ph.Eur./BP
- Cl (Chloride) [Ph.Eur./BP]:≤ 300 ppm
- Fe (Iron) [Ph.Eur./BP]:≤ 20 ppm
- Loss on drying (400°C) [Ph.Eur./BP]:48.0 - 52.0 %
- USP specifications::.
- Assay (by ignition) [USP]:98.0 - 102.0 %
- Cl (Chloride) [USP]:≤ 0.014 %
- Fe (Iron) [USP]:≤ 20 μg/g
- Se (Selenium) [USP]:≤ 30 μg/g
- pH (25°C; 50 mg/ml) [USP]:5.0 - 9.2
- Loss on ignition (450 ± 25°C) [USP]:40.0 - 52.0 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMagnesium Sulphate Heptahydrate Ph. Eur., USP, BP
Additional Product Details
sds_VWRC6527
SDS
spec_VWRC6527.1000 spec_VWRC6527.1000 spec_VWRC6527.1000 spec_VWRC6527.5000 spec_VWRC6527.5000
Product specification
More
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together