Order Entry
India
ContactUsLinkComponent
Mercury(II) sulphate ≥97%, purified
Toxic
Health
Environment
Mercury(II) sulphate ≥97%, purified
Catalog # 25445.233
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Health
Environment
Mercury(II) sulphate ≥97%, purified
Catalog # 25445.233
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

 :
 :296.65 g/mol
Merck Index:13,05914
Melting Pt:850 °C
Density:6,47 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-992-5
UN:1645
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
CAS Number:7783-35-9
Formula:HgSO₄
MDL Number:MFCD00011047