Order Entry
India
ContactUsLinkComponent
 
 
 
 
Imidazole ≥99%, Proteomics Grade
  M136-100G
 :  
undefined
 
 
 
Imidazole ≥99%, Proteomics Grade
  M136-100G
 :  

 

  • Pack type:
    Bottle, amber
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • DNase:
    none detected
  • Appearance:
    White to slightly yellow solid
  • Insolubles:
    ≤0.1%
  • Melting Point:
    88 – 92 °C
  • pH (5%, Water) @25 °C:
    9.5 – 10.5
  • Protease:
    none detected
  • Purity:
    ≥99%
  • RNase:
    none detected
  • Solubility (20%, Water):
    Pass

 

 

 :
 :13,04935
 :255 °C (1013 hPa)
 :89…90 °C
 :1,18 g/cm³ (20 °C)
 :135 °C
 :Ambient
 :206-019-2
 :3263
 :8,II
 :68.08 g/mol
 :GHS08
 :GHS07
 :GHS05
 :288-32-4
 :C₃H₄N₂
 :MFCD00005183