Ammonium sulphate
Catalog # A3485.5000
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
About this item
:
:132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1,78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-984-1
REACH:01-2119455044-46
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391