
Ammonium hydrogen carbonate ≥97.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 21219.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 97.5 %
- Heavy metals (as Pb):Max. 5 ppm
- Ignition residue (SO4):Max. 50 ppm
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 20 ppm
- Fe (Iron):Max. 2 ppm
Specification Test Results
About this item
:
:79.06 g/mol
Merck Index:13,00497
Melting Pt:35 °C
Density:1.57 g/cm³ (20 °C)
EINECS:213-911-5
:GHS07
CAS Number:1066-33-7
Formula:NH₄HCO₃
MDL Number:MFCD00012138
Density:1,57 g/cm³ (20 °C)