Order Entry
India
ContactUsLinkComponent
Cetrimonium bromide
Cetrimonium bromide
Catalog # A0805.0500
Supplier:  PanReac AppliChem
CAS Number:  
Cetrimonium bromide
Catalog # A0805.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :364.45 g/mol
Merck Index:13,02034
Melting Pt:237…243 °C
Flash Pt:244 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-311-3
UN:3077
ADR:9,III
 :GHS08
 :GHS07
 :GHS05
 :GHS09
CAS Number:57-09-0
Formula:CH₃(CH₂)₁₅N(Br)(CH₃)₃
MDL Number:MFCD00011772