Order Entry
India
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Sodium sulphate, anhydrous ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 99.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 0.01 %
  • ACS - Assay (Na₂SO₄):
    ≥ 99.0 %
  • ACS - Insoluble Matter:
    ≤ 0.01 %
  • ACS - Loss on Ignition:
    ≤ 0.5 %
  • ACS - pH of 5% Solution at 25°C:
    5.2 - 9.2
  • ACS - Chloride (Cl):
    ≤ 0.001 %
  • ACS - Phosphate (PO₄):
    ≤ 0.001 %
  • ACS - Calcium (Ca):
    ≤ 0.01 %
  • ACS - Magnesium (Mg):
    ≤ 0.005 %
  • ACS - Potassium (K):
    ≤ 0.01 %
  • ACS - Nitrogen Compounds (as N):
    ≤ 5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • ACS - Iron (Fe):
    ≤ 0.001 %

Specification Test Results

Related Information

Frequently Bought Together