


Potassium peroxodisulphate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7727-21-1
:
Formula:K₂S₂O₈
MDL Number:MFCD00011386
:270.32 g/mol
Merck Index:13,07741
Melting Pt:100 °C
Density:2,477 g/cm³ (20 °C)
EINECS:231-781-8
UN:1492
ADR:5.1,III
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 98.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Insolubility in water:Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- Mn (Manganese):Max. 2 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
spec_VWRC26915.187 spec_VWRC26915.187 spec_VWRC26915.291
Product specification
Frequently Bought Together