Order Entry
India
Orders LinkContactUsLinkComponent
 

Water

   Sustainable Options Available
Supplier:   BDH US PRODUCED
Water
Supplier:  BDH US PRODUCED
   Sustainable Options Available
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • BDHB1168-4LP
  • BDHB1168-5GL
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Environmental certification:
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Product Family Options

Product Information

  • Pk Info
    Pk
    Availability
    Price
  • 4x4 L
    4
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    BDHB1168-4LP
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • 19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L

    Specifications:
    More
    Specifications
    Cat. No.
    BDHB1168-5GL
    Environmentally Preferable
    Environmental certification
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.