Order Entry
India
ContactUsLinkComponent
Thymol ≥99%, GPR RECTAPUR®
Thymol ≥99%, GPR RECTAPUR®
Catalog # 20728.237
Supplier:  VWR Chemicals
CAS Number:  
undefined
Thymol ≥99%, GPR RECTAPUR®
Catalog # 20728.237
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Melting point:
    48 - 52 °C
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :150.22 g/mol
Merck Index:13,09476
Boiling Pt:233 °C (1013 hPa)
Melting Pt:49…51 °C
Density:0,965 g/cm³ (25 °C)
Flash Pt:104 °C
EINECS:201-944-8
UN:2430
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:89-83-8
Formula:2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
MDL Number:MFCD00002309