
Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:532-32-1
:
Formula:C₆H₅COONa
MDL Number:MFCD00012463
:144.11 g/mol
Merck Index:13,08654
Melting Pt:410…430 °C
Density:1,44 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Flash Pt:>100 °C
EINECS:208-534-8
REACH:01-2119460683-35
:GHS07
Storage Temperature:Ambient
Specification Test Results
- GMP Manufactured Product:—
- Meets E.P. Chemical Specifications:—
- Meets N.F. Requirements:—
- Meets B.P. Chemical Specifications:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Identification C:Passes Test
- NF - Alkalinity:Passes Test
- NF - Water (H₂O):≤ 1.5 %
- NF - Assay (C₆H₅COONa) (anhydrous basis):99.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Halogenated Compounds (as Cl) - Ionized Chlorine:≤ 200 ppm
- EP/BP - Halogenated Compounds (as Cl) - Total Chlorine:≤ 300 ppm
- EP/BP - Loss on Drying:≤ 2.0 %
- EP/BP - Assay(C₆H₅COONa)(dried basis):99.0 - 100.5 %
- No Class 1,2,3 or other solvents are used or produced in the:99.0 - 100.5 %
Specification Test Results
Frequently Bought Together