Order Entry
India
ContactUsLinkComponent
Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, FCC, J.T.Baker®
Irritant
Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, FCC, J.T.Baker®
Catalog # 3500-05
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Sodium benzoate 99.0-101.0% (by anhydrous basis) NF, FCC, J.T.Baker®
Catalog # 3500-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Bulk Food Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Identification C:
    Passes Test
  • NF - Alkalinity:
    Passes Test
  • NF - Water (H₂O):
    ≤1.5 %
  • NF - Assay (C₆H₅COONa) (anhydrous basis):
    99.0 - 101.0 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Identification C:
    Passes Test
  • FCC - Alkalinity:
    ≤0.04 %
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Water (H₂O):
    ≤1.5 %
  • FCC - Assay (C₆H₅COONa) (anhydrous basis):
    99.0 - 101.0 %

Specification Test Results

About this item

 :
 :144.11 g/mol
Merck Index:13,08654
Melting Pt:410…430 °C
Density:1,44 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:208-534-8
REACH:01-2119460683-35
 :GHS07
Storage Temperature:Ambient
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463