

tert-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Catalog # 20814.292
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity:Max. 0.0002 meq/g
- Boiling point:81.5 - 83.0 °C
- Colouration:Max. 10 APHA
- Density (26/4):0.770 - 0.780
- Solidification point:24 - 26 °C
- Non-volatile residue:Max. 20 ppm
- Water:Max. 0.1 %
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,01577
Boiling Pt:82,2 °C (1013 hPa)
Melting Pt:25,3 °C
Density:0,7887 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-889-7
UN:1120
ADR:3,II
REACH:01-2119444321-51
:GHS02
:GHS07
CAS Number:75-65-0
Formula:(CH₃)₃COH
MDL Number:MFCD00004464