J.T.Baker® BAKERBOND Speedisk® Hydrophilic DVB, SPE Columns
: Avantor
These columns feature BAKERBOND® DVB polymer resins, which are the product of ultra-clean polymer microparticle technology. These resin particles have a large surface area, are highly rigid, are stable over pH range 1 to 14, are water-wettable and are not impacted by sorbent drying. These columns are recommended when advanced detection methods are used.
- Improve sample preparation recovery
- Polymer based
General applications: Polar to non polar analytes.
These SPE columns can be used in adsorption mode.
Frequently Bought Together