



2-Mercaptoethanol ≥99%, Proteomics Grade
Catalog # M131-250ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Bottle, amber
- Pk:250 mL
- Seq:2590
Specifications
Specification Test Results
- DNase:none detected
- Protease:none detected
- Purity:≥99%
- RNase:none detected
- Water:≤0.5%
Specification Test Results
About this item
:
Merck Index:13,05894
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–100 °C
Density:1,12 g/cm³ (20 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:200-464-6
UN:2966
ADR:6.1,II
:78.14 g/mol
:GHS06
:GHS08
:GHS05
:GHS07
CAS Number:60-24-2
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890