Order Entry
India
ContactUsLinkComponent
 
Succinic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
  20668.230
 :  
undefined
Succinic acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
  20668.230
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Assay:
    Min. 99.5 %
  • Melting point:
    185-191 °C
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.02 %
  • Insolubility in water:
    Max. 50 ppm
  • Water:
    Max. 0.5 %
  • Cl (Chloride):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm

 

 

 :
 :118.09 g/mol
 :13,08953
 :235 °C (1013 hPa)
 :182 °C
 :1.56 g/cm³ (20 °C)
 :1,56 g/cm³ (20 °C)
 :206 °C
 :Ambient
 :203-740-4
 :3261
 :8,III
 :GHS07
 :110-15-6
 :HOOCCH₂CH₂COOH
 :MFCD00002789