Dichloromethane ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Methylene chloride
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
Merck Index:12,06140
Boiling Pt:39,8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1,322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
ADR:6.1,III
:84.93 g/mol
UN:1593
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (by GC, corrected for water, exclusive of preservative):≥99.8 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤233
- Residue after Evaporation:≤2.0 ppm
- Titrable Acid (meq/g):≤0.0003
- Chloride (Cl):≤10 ppm
- Water (by KF, coulometric):≤0.010 %
- Preservative (cyclohexene):≥50 ppm
Specification Test Results
Frequently Bought Together