Order Entry
India
ContactUsLinkComponent
L(+)-Isoleucine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
L(+)-Isoleucine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Catalog # 2082-05
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Isoleucine 98.5-101.5% USP, Multi-Compendial, J.T.Baker®
Catalog # 2082-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • USP - Assay (C₆H₁₃NO₂) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation α²⁵D (+):
    38.9 - 41.8 Degree
  • USP - pH:
    5.5 - 7.0
  • USP - Loss on Drying at 105°C:
    ≤ 0.3 %
  • USP - Residue on Ignition:
    ≤ 0.3 %
  • USP - Chloride (Cl):
    ≤ 0.05 %
  • USP - Sulfate (SO₄):
    ≤ 0.03 %
  • USP - Iron (Fe):
    ≤ 0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤ 0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤ 2.0 %
  • EP/BP - Assay (C₆H₁₃NO₂) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotationα²ºD (+):
    40.0 - 43.0 Degree
  • EP/BP-Ninhydrin-Positive Substances-Impurity A:
    ≤ 0.3 %
  • EP/BP-Ninhydrin-Positive Substances-Impurity C:
    ≤ 0.3 %
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤ 0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤ 1.0 %
  • EP/BP - Chloride (Cl):
    ≤ 200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤ 0.02 %
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JP - Assay (C₆H₁₃NO₂) (dried basis):
    98.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation (+):
    39.5 - 41.5 Degree
  • JP - pH:
    5.5 - 6.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.021 %
  • JP - Sulfate (SO₄):
    ≤ 0.028 %
  • JP - Ammonium (NH₄):
    ≤ 0.02 %
  • JP - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤ 0.30 %
  • JP - Residue on Ignition:
    ≤ 0.10 %
  • Endotoxin Concentration, IU/mg, For Information Only:
    = 0.028 %
  • Only Class 2 Solvents (Methanol) are likely to be present. All are below Option 1 limits.:
    = 0.02 %
  • Elemental Impurities (USP (232), EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:
    = 20 ppm
  • Endotoxin Concentration, IU/mg:
    For Information Only

Specification Test Results

About this item

 :
 :131.17 g/mol
Merck Index:14,05179
Melting Pt:279…280 °C
Storage Temperature:Ambient
EINECS:200-798-2
CAS Number:73-32-5
Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
MDL Number:MFCD00064222