Order Entry
India
ContactUsLinkComponent
L(+)-Tartaric acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Corrosive
L(+)-Tartaric acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
L(+)-Tartaric acid ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Insolubility in water:
    Max. 50 ppm
  • C2O4 (Oxalate):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 5 ppm
  • PO4 (Phosphate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Fe (Iron):
    Max. 5 ppm

Specification Test Results

Frequently Bought Together