L(+)-Glutamine 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
: Avantor
:56-85-9
:
:C₅H₁₀N₂O₃
:MFCD00008044
:146.15 g/mol
:13,04484
:355 °C (1013 hPa)
:185…186 °C
:1,5326 g/cm³ (130 °C)
:Management of Change (MOC) category = R
:Ambient
:200-292-1
- USP - Assay (C₅H₁₀N₂O₃) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²º^D (+):6.3 - 7.3 Degree
- USP - Loss on Drying at 105°C:≤ 0.3 %
- USP - Residue after Ignition:≤ 0.3 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds:0.5% max
- JP - Assay (C₅H₁₀N₂O₃) (dried basis):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Specific Rotation [Å]²º^D (+):6.3 - 7.3 Degree
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP - Residue after Ignition:≤ 0.1 %
- JP - Chloride (Cl):≤ 0.021 %
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Iron (Fe):≤ 10 ppm
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Related substances:Passes Test
- JP - pH:4.5 - 6.0
- JP - Clarity and Color of Solution:Passes Test
- JP - Ammonium:≤ 0.1 %
- Solubility of 3% Solution at 30°C:Passes Test
- Solubility of 3.5% Solution at 30°C:Passes Test
- Identification (by IR):Passes Test
- Endotoxin Concentration (EU/mg):≤ 0.06
- Endotoxin Concentration, IU/mg:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:Passes Test
- manufacturing or purification of the product.:= 0.3 %
- FCC - Specific Rotation [Å]²⁰ D (+):6.3 - 7.3 Degree
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental:Passes Test
- impurities for this product is available on the associated Product:Passes Test
- Regulatory Data Sheet and elemental impurity profile report.:Passes Test
Frequently Bought Together