Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
Rs.0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
India
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
VWR®, Organic reference standard, Chloroethylene 200 µg/ml in methanol
VWR®, Organic reference standard, Chloroethylene 200 µg/ml in methanol
VWR®, Organic reference standard, Chloroethylene 200 µg/ml in methanol
123462Y
:
VWR Chemicals
:
VWR®, Organic reference standard, Chloroethylene 200 µg/ml in methanol
123462Y
:
VWR Chemicals
:
Pack type:
Glass ampoule
Pk:
1 mL
CAS No.:
75-01-4
Description:
Chloroethylene 200 µg/ml in methanol
Formula:
C₂H₃Cl
Description:
Chloroethylene