Order Entry
India
ContactUsLinkComponent
Iron(II) chloride tetrahydrate ≥99%, purified
Irritant
Corrosive
Iron(II) chloride tetrahydrate ≥99%, purified
Catalog # 24128.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
Iron(II) chloride tetrahydrate ≥99%, purified
Catalog # 24128.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

About this item

 :
Melting Pt:677 °C
Density:1,926 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,III
 :198.81 g/mol
EINECS:231-843-4
 :GHS07
 :GHS05
CAS Number:13478-10-9
Formula:FeCl₂·4H₂O
MDL Number:MFCD00149709