Forensic DNA kit, E.Z.N.A.®
E.Z.N.A.® Forensic DNA Isolation Kit, 5 Tests
Catalog # D3591-00
Supplier: OMEGA BIOTEK
Jump to:
Product Details & Documents
The E.Z.N.A.® Forensic DNA Kit is designed to provide a rapid and easy method for the isolation of genomic DNA from forensic samples such as dry blood, buccal swabs, and semen samples for consistent PCR and Southern analysis. This kit can also be used for the preparation of genomic DNA from mouse tail snips, whole blood, buffy coat, serum, and plasma.
- Process single or multiple samples simultaneously
- No need for phenol/chloroform extractions
- Time-consuming steps such as precipitation with isopropanol or ethanol are eliminated
DNA purified using the E.Z.N.A.® Forensic DNA Kit is ready for applications such as PCR, Southern blotting, and restriction enzyme digestion.
Sample size: 30 mg tissue or 107 cells
Specifications
Specifications
- Pk:5 Tests
- Description:E.Z.N.A.® Forensic DNA Isolation Kit
- Cat. No.:D3591-00
- Prep size:Mini
- Refine target molecule:Total DNA
- Target molecule:DNA
- Method / Format:Column (silica-based)
- Description:E.Z.N.A.® Forensic DNA Isolation Kit
- Sample type:Forensic samples



