Copper(II) chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10125-13-0
:
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674
:170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C
Density:2,54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS07
:GHS09
Specification Test Results
- Assay:Min. 99.0 %
- pH (20°C; 5 %):3.0 - 3.8
- Insolubility in water:Max. 100 ppm
- Total N (Nitrogen):Max. 30 ppm
- SO4 (Sulphate):Max. 50 ppm
- As (Arsenic):Max. 1 ppm
- Fe (Iron):Max. 30 ppm
- Ni (Nickel):Max. 10 ppm
- Pb (Lead):Max. 40 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
23093_COPPER (II) CHLORIDE 2H2O ANALAR NP R.PE_BSE_REV0
BSE/TSE Certificates
spec_VWRC23093.233 spec_VWRC23093.233 spec_VWRC23093.290 spec_VWRC23093.290
Product specification
Frequently Bought Together