Magnesium chloride hexahydrate 99.0-102.0%, crystals, Ultrapure Bioreagent for molecular biology, J.T.Baker®
BAKR4003-01
: Avantor
:
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
- For Laboratory,Research,or Manufacturing Use:99.0 - 102.0 %
- Assay (MgCl₂·6H₂O):99.0 - 102.0 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- Insoluble Matter:≤ 0.005 %
- Ammonium (NH₄):≤ 0.002 %
- Calcium (Ca):≤ 0.01 %
- Potassium (K):≤ 0.005 %
- Sodium (Na):≤ 0.005 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤ 5 ppm
- Trace Impurities - Iron (Fe):≤ 5 ppm
- Trace Impurities - Manganese (Mn):≤ 5.0 ppm
: Magnesium dichloride
:
:203.3 g/mol
:13,05684
:1412 °C (1013 hPa)
:117 °C
:1,57 g/cm³ (25 °C)
:Ambient
:232-094-6
:7791-18-6
:MgCl₂·6H₂O
:MFCD00149781