Order Entry
India
ContactUsLinkComponent
Disodium hydrogen phosphate dihydrate Ph. Eur., USP
Disodium hydrogen phosphate dihydrate Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates
undefined
Disodium hydrogen phosphate dihydrate Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Heavy metals (as Pb):
    ≤ 20 ppm
  • Residual solvents:
    Unlikely by manuf.process
  • Residues of metal catalysts or reagents:
    Unlikely by manuf.process
  • As (Arsenic):
    ≤ 2 ppm
  • Cd (Cadmium):
    ≤ 1 ppm
  • Co (Cobalt):
    ≤ 1 ppm
  • Hg (Mercury):
    ≤ 2 ppm
  • Ni (Nickel):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 2 ppm
  • V (Vanadium):
    ≤ 3 ppm
  • Conforms to Ph.Eur./USP:
    Passes test
  • Ph.Eur. specifications::
    .
  • Assay (dried substance) [Ph.Eur.]:
    98.0 - 101.0 %
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Identification C:
    Passes test Ph.Eur.
  • Identification D:
    Passes test Ph.Eur.
  • Appearance of solution:
    Passes test Ph.Eur.
  • Reducing substances:
    Passes test Ph.Eur.
  • Monosodium phosphate [Ph.Eur.]:
    ≤ 2.5 %
  • Cl (Chloride) [Ph.Eur.]:
    ≤ 400 ppm
  • SO4 (Sulphate) [Ph.Eur.]:
    ≤ 0.1 %
  • Fe (Iron) [Ph.Eur.]:
    ≤ 40 ppm
  • Loss on drying (130°C) [Ph.Eur.]:
    19.5 - 21.0 %
  • USP specifications::
    .
  • Assay (calculated on dried basis) [USP]:
    98.0 - 100.5 %
  • Insoluble substances [USP]:
    ≤ 0.4 %
  • Cl (Chloride) [USP]:
    ≤ 0.06 %
  • SO4 (Sulphate) [USP]:
    ≤ 0.2 %
  • As (Arsenic) [USP]:
    ≤ 16 ppm
  • Loss on drying (130°C) [USP]:
    18.5 - 21.5 %

Specification Test Results

Frequently Bought Together