
Potassium hydroxide concentrate for dilution to 1 mol/l (1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4674-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:LDPE ampoule
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.998-1.002
- Normality (by titrimetry):Passes Test
Specification Test Results
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,III
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553