Order Entry
India
ContactUsLinkComponent
 
Irritant
Corrosive
DL-Malic acid ≥99%, TECHNICAL
Catalog # 20365.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Corrosive
DL-Malic acid ≥99%, TECHNICAL
Catalog # 20365.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %

Specification Test Results

About this item

 :
 :134.09 g/mol
Merck Index:13,05730
Boiling Pt:306 °C (1013 hPa)
Melting Pt:131…132 °C
Density:1,601 g/cm³ (20 °C)
EINECS:230-022-8
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:617-48-1
Formula:HOOCCH(OH)CH₂COOH
MDL Number:MFCD00064212