Order Entry
India
ContactUsLinkComponent
 
VWR®, Gelatin
 
undefined
VWR®, Gelatin
 

 

  • CAS No.:
    9000-70-8
  • Storage conditions:
    Ambient
  • Protein synonyms:
    Gelatine
  • Protein/peptide name:
    Gelatin
  • Grade:
    Reagent Grade

 

 

Frequently Bought Together