Order Entry
India
ContactUsLinkComponent
Petroleum spirit, 100…140 °C, BAKER, Laboratory Reagent, J.T.Baker®
Flammable
Health
Irritant
Environment
Petroleum spirit, 100…140 °C, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8243.1000
Supplier:  Avantor
CAS Number:  
Flammable
Health
Irritant
Environment
Petroleum spirit, 100…140 °C, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 8243.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Boiling Range:
    100 - 140 °C
  • Residue after Evaporation:
    max. 0.005 %

Specification Test Results

About this item

 :
Boiling Pt:100…140 °C (1013 hPa)
Melting Pt:–40 °C
Density:0,728 g/cm³ (15 °C)
Flash Pt:1 °C
Storage Temperature:Ambient
EINECS:265-151-9
UN:1268
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849