VWR®, Filter tips, Standard Line
VWRU10017-100
- Packed:96/rack, 10 racks/pack
- Sterile:+
- Volume:200 µl
- Pk:960
VWR® Standard Line pipette tips utilise the industry standard in aerosol barrier technology.
- Translucent packaging for easy vizualization of tips
- Built with stability for ease of use when multi-channel pipetting
- Back hinge makes it easy to open and close with one hand
- Certified free of contaminants for the most sensitive laboratory protocols
- Conveniently racked in the most widely used style of rack on the market