Order Entry
India
ContactUsLinkComponent
VWR®, Seals and Closures for 11 mm Crimp Top Vials
VWR®, Seals and Closures for 11 mm Crimp Top Vials
Catalog # VWRU46610-744
Supplier:  VWR Collection
VWR®, Seals and Closures for 11 mm Crimp Top Vials
Catalog # VWRU46610-744
Supplier:  VWR Collection
Specifications
  • Septum:
    Clear PTFE/natural red rubber
  • Description:
    Aluminum seal
  • Seq:
    02
  • Pk:
    100