Order Entry
India
ContactUsLinkComponent
VWR®, Meker Burners
VWR®, Meker Burners
Catalog # VWRU89038-544
Supplier:  VWR Collection
undefined
VWR®, Meker Burners
Catalog # VWRU89038-544
Supplier:  VWR Collection
Specifications
  • Description:
    Meker burner
  • Seq:
    001
  • Pk:
    1
  • For:
    Natural gas
Specifications
About this item
Meker-type high-temperature burners are for use where steady high temperatures are required.
  • Backflashes eliminated via safety cap feature
  • Heavy die-cast base
  • Uniform, consistent temperature
The 86 mm Meker grid top forms short, uniform blue cones of consistent temperature. Safety cap design eliminates backflashes. Heavy die-cast base unit and venturi tube are made from nickel-plated zinc alloy.
Grid dimension (Ø×H): 38×216 mm.