Order Entry
India
ContactUsLinkComponent
VWR®sympHony™, Multi-parameter meters, bench top
undefined
VWR®sympHony™, Multi-parameter meters, bench top
  • Catalog No:
  • VWRU89231-698
  • VWRU89231-664
  • VWRU89231-692
  • Model:
  • B10C
  • pH range:
  • −2...+19
  • pH resolution:
  • 0,1/0,01/0,001
  • pH accuracy:
  • ≤0,002 (±1 digit)
  • ISE range:
  • -
  • Conductivity range:
  • -
  • Conductivity resolution:
  • -
  • Conductivity accuracy:
  • -
  • Resistivity range:
  • -
  • Resistivity accuracy:
  • ≤0,5% (±1 digit)
  • Salinity range:
  • -
  • DO concentration:
  • -
  • DO concentration resolution:
  • -
  • DO accuracy:
  • -
Frequently Bought Together