Order Entry
India
ContactUsLinkComponent
VWR®, Hazardous Location/Explosion Proof Laboratory Refrigerators and Freezers
VWR®, Hazardous Location/Explosion Proof Laboratory Refrigerators and Freezers
Catalog # VWRU97055-760
Supplier:  VWR Collection
undefined
VWR®, Hazardous Location/Explosion Proof Laboratory Refrigerators and Freezers
Catalog # VWRU97055-760
Supplier:  VWR Collection

Specifications

  • W×D×H ext.:
    864×895×1988 mm
  • Capacity:
    566 l
  • Description:
    Explosion proof hazardous location refrigerator, EPR-2004
  • Seq:
    06
  • Model:
    EPR-2004
  • Cat. No.:
    VWRU97055-760
  • Shelves/drawers:
    1 adjustable shelves, 3 fixed shelves
  • Weight:
    144 kg
  • Pk:
    1
  • Temperature:
    0…+10 °C

Specifications

About this item

In these units, all sources of sparks and ignition in these refrigerators and freezers such as lights, thermostats, switches, fans, relays, and compressors have been removed or sealed to accommodate storage of flammable materials in a hazardous area.

  • Designed for the storage of flammable materials
  • For use in hazardous areas
  • Units meet NFPA and OSHA guidelines
  • Two-year parts and labor warranty
  • Three-year warranty on compressor parts

Units have exterior adjustable mechanical thermostats. The exteriors feature a white baked-on powder coat finish, and single solid doors with keyed locks.

Certifications: Units meet NFPA and OSHA guidelines and are UL and C-UL listed.
Delivery information: These units are not supplied with a power cord, and are required to be hard wired in a conduit to a 120 VAC power source.