
DSG (Disuccinimidyl glutarate) ≥99%, BioChemica®
Catalog # A7822.0001
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- State of matter:Solid
- Storage:-20 °C
- Assay:min. 99%
- Melting point:142 - 145 °C
- Identity (NMR):Passes test
Specification Test Results
About this item
:
:326.26 g/mol
Melting Pt:142…145 °C
:GHS07
Storage Temperature:Freezer
CAS Number:79642-50-5
Formula:C₁₃H₁₄N₂O₈
MDL Number:MFCD00153597