Rs.0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
India
ContactUsLinkComponent
Centrifuges
Rotors
VWR®, Rotor for Compact VWR® Centrifuge
VWR®, Rotor for Compact VWR® Centrifuge
VWRU10799-172
:
VWR Collection
VWR®, Rotor for Compact VWR® Centrifuge
VWRU10799-172
:
VWR Collection
Description:
24-place microtube rotor, 40°, 15000 min
-1
, 21382 ×g
Pk:
1