VWR® ECO Funnel™, Safety funnel system
VWRMNPEF3004CSYS
: VWR USA
- Ø:203 mm
- Capacity:1750 ml
- Description:Complete funnel system with 4 l bottle and secondary container
- Seq:01
- Pk:1
The ECO Funnel™ system is a key safety product designed to reduce exposure to hazardous waste and to prevent toxic chemical evaporation.
- Durable, chemical resistant HDPE construction
- 2000 ml capacity ECO Funnel™ with containers available from 4 to 208 litres
- Built-in liquid level indicator tube helps prevent the container from over-filling
- Comes with secure lid, positive latch, gasket, filter, 11" long stem to reduce chemical evaporation, and industrial strength screw cap for common laboratory containers
Funnels can be accessorised to connect to HPLC waste line, or solvents can be poured directly into the funnel. They have a custom-designed air tube vent that allows solvent waste to flow easily and quickly during disposal.
: Complete system includes 8" ECO Funnel™, waste bottle or container, and secondary container with or without base (in most cases). All ECO funnels are available with a red or yellow lid (specify lid colour when ordering).