Order Entry
India
Orders LinkContactUsLinkComponent
Buffer, Tris-EDTA solution
Tris-EDTA solution, clear, liquid, 1 L
  BDHB7732-1
Buffer, Tris-EDTA solution
  BDHB7732-1
 :  
Buffer, Tris-EDTA solution
Tris-EDTA solution, clear, liquid, 1 L
 :  
1 * 1 L
Tris-EDTA solution (TE solution) contains ultra pure deionised water, 0,5 M EDTA with pH 7,5 and 1 M tris with pH 8.
  • Pk:
    1 L
  • Description:
    Tris-EDTA solution, clear, liquid