Order Entry
India
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis) USP, J.T.Baker®
Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis) USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤16 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.06%
  • Bulk Pharmaceutical Chemical:
    ≤2.5
  • Appearance (Colorless or translucent crystals or granules):
    Passes Test
  • USP - Assay (Na₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤ 16 ppm
  • USP - Chloride (Cl):
    ≤ 0.06 %
  • USP - Endotoxin Concentration (EU/g):
    ≤ 2.5
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.4 %
  • USP - Loss on Drying:
    43.0 - 50.0 %
  • USP - Sulfate (SO₄):
    ≤ 0.2 %
  • EP - Identification A:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Identification D:
    Passes Test
  • EP - Reducing Substances:
    Passes Test
  • EP - Monosodium Phosphate:
    ≤ 2.5 %
  • EP - Chloride (Cl):
    ≤ 200 ppm
  • EP - Sulfate (SO₄):
    ≤ 500 ppm
  • EP - Appearance of Solution:
    Passes Test
  • EP - Iron (Fe):
    ≤ 20 ppm
  • EP - Assay (dried basis):
    98.0 - 101.0 %
  • JP - Arsenic (As):
    ≤ 2 ppm
  • JP - Assay (Na₂HPO₄) (dried basis):
    98.0 - 101.0 %
  • JP - Carbonate (CO₃):
    Passes Test
  • JP - Chloride (Cl):
    ≤ 0.014 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals:
    ≤ 10 ppm
  • JP - Identification:
    Passes Test
  • JP - pH (1 in 50):
    9.0 - 9.4
  • JP - Sulfate (SO₄):
    ≤ 0.038 %

Specification Test Results

Frequently Bought Together