(±)-1,2-Propanediol ≥99.5% USP, FCC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:57-55-6
:
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272
:76.1 g/mol
Merck Index:13,07947
Boiling Pt:187,6 °C (1013 hPa)
Melting Pt:–59 °C
Density:1,04 g/cm³ (20 °C)
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:200-338-0
Specification Test Results
- Meets U.S.P Requirements:—
- GMP Manufactured Product:—
- Food GMP Manufactured Product:—
- Meets F.C.C. Requirements:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- Bulk Food Chemical:—
- USP - Specific Gravity at 25°/25°C:1.035 - 1.037
- USP - Acidity:Passes Test
- USP - Water (H₂O)(by Karl Fischer titrn):≤0.2 %
- USP - Identification A (IR):Passes Test
- USP - Identification B - Diethylene Glycol:≤0.10 %
- USP - Identification B - Ethylene Glycol:≤0.10 %
- USP - Identification C:Passes Test
- USP - Residue on Ignition:≤0.007 %
- USP - Chloride (Cl):≤0.007 %
- USP - Sulfate (SO₄):≤0.006 %
- USP - Assay (CH₃CHOHCH₂OH):≥99.5 %
- FCC - Specific Gravity at 25°/25°C:1.035 - 1.037
- FCC - Water (H₂O)(by Karl Fischer titrn):≤0.20 %
- FCC - Identification A (IR):Passes Test
- FCC - Lead (Pb):≤1 mg/kg
- FCC - Residue on Ignition:≤0.007 %
- FCC - Assay (CH₃CHOHCH₂OH):≥99.5 %
- FCC - Distilling Range(Lower):185 - 189 °C
- FCC - Distilling Range(Upper):185 - 189 °C
- FCC - Acidity:Passes Test